Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: fynb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001029969.1 | NP_001025140.1 | 9 - 10 | 1264 | 68 | |
XM_005160302.4 | XP_005160359.1 | 9 - 10 | 1311 | 68 | ||
XM_009294487.3 | XP_009292762.1 | 8 - 9 | 1146 | 68 | ||
XM_021468512.1 | XP_021324187.1 | 8 - 9 | 1253 | 68 | ||
GenBank mRNA | BC098534.1 | - | 9 - 10 | 1264 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |