Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ftr67 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001039719.2 | NP_001034808.2 | 3 - 4 | 994 | 79 | |
GenBank mRNA | BC076205.1 | - | 3 - 4 | 79 | ||
BC093187.1 | - | 3 - 4 | 79 | |||
BC114247.1 | - | 3 - 4 | 79 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Danio rerio Probe spans exons |