Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: cldn15b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001040314.1 | NP_001035404.1 | 3 - 4 | 629 | 69 | |
GenBank mRNA | BC115253.1 | - | 3 - 4 | 629 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Danio rerio Probe spans exons |