Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: pdlim2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001326402.1 | NP_001313331.1 | 5 - 6 | 640 | 120 | |
XM_009304150.3 | XP_009302425.1 | 5 - 6 | 770 | 120 | ||
GenBank mRNA | BC139715.1 | - | 5 - 6 | 541 | 120 | |
DQ679230.2 | - | 5 - 6 | 545 | 120 | ||
EB977268.1 | - | 5 - 6 | 592 | 120 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Danio rerio Probe spans exons |