Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: rfng | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001001830.2 | NP_001001830.2 | 5 - 6 | 1183 | 61 | |
GenBank mRNA | AF510993.1 | - | 5 - 6 | 61 | ||
BC118683.1 | - | 5 - 6 | 61 | |||
BC139600.1 | - | 5 - 6 | 1185 | 61 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Danio rerio Probe spans exons |