Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: olfml3a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001045857.1 | NP_001039322.1 | 2 - 3 | 523 | 142 | |
GenBank mRNA | BC122413.1 | - | 2 - 3 | 523 | 142 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Danio rerio Probe spans exons |