Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: nsrp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001077430.2 | NP_001070898.1 | 5 - 6 | 523 | 117 | |
GenBank mRNA | BC076130.1 | - | 5 - 6 | 543 | 117 | |
BC092757.1 | - | 5 - 6 | 524 | 117 | ||
BC110090.1 | - | 5 - 6 | 535 | 117 | ||
BC124124.1 | - | 5 - 6 | 528 | 117 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |