Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: crfb2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001077626.2 | NP_001071094.2 | 2 - 3 | 401 | 121 | |
XM_005167960.4 | XP_005168017.1 | 2 - 3 | 496 | 121 | ||
GenBank mRNA | EF014953.1 | - | 2 - 3 | 401 | 121 |
Set Membership: |
Amplicon length greater than or equal to 101 Danio rerio Probe spans exons |