Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
Home
›
Search Tool
›
Search Results
› Dr03073267_m1
See other ELNA GE Assays ›
Gene Symbol:
elna
Gene Name:
elastin a
Gene Aliases:
eln1
Chromosome Location:
Chr.15: 263189 - 323096 on Build GRCz11
Species:
Zebrafish
Species Specific ID (Flybase ID):
-
Assay ID
Dr03073267_m1
Size
FAM-MGB | XS: 75 rxns
FAM-MGB | S: 360 rxns
FAM-MGB | M: 750 rxns
FAM-MGB | L: 2900 rxns
VIC-MGB_PL | S: 360 rxns
VIC-MGB_PL | M: 750 rxns
VIC-MGB_PL | L: 2900 rxns
VIC-MGB | S: 360 rxns
VIC-MGB | M: 750 rxns
VIC-MGB | L: 2900 rxns
Availability
Made To Order
Catalog #
4351372
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
Assay Details
More Information
Genomic Map
LOADING...
×
Back To Top
Assay Details
Gene Symbol: elna
Interrogated Sequence
Translated Protein
Exon Boundary
Assay Location
IMAGE Clone ID
Amplicon Length
RefSeq
NM_001080063.2
NP_001073532.2
51 - 52
3289
69
XM_009291337.3
XP_009289612.1
50 - 51
3504
69
XM_017351021.2
XP_017206510.1
52 - 53
3694
69
XM_017351022.2
XP_017206511.1
52 - 53
3689
69
XM_017351023.2
XP_017206512.1
52 - 53
3689
69
XM_017351024.2
XP_017206513.1
52 - 53
3689
69
XM_017351025.2
XP_017206514.1
52 - 53
3682
69
XM_017351026.2
XP_017206515.1
52 - 53
3679
69
XM_017351027.2
XP_017206516.1
52 - 53
3670
69
XM_017351028.2
XP_017206517.1
51 - 52
3651
69
XM_017351029.2
XP_017206518.1
51 - 52
3649
69
XM_017351031.2
XP_017206520.1
50 - 51
3566
69
XM_017351032.2
XP_017206521.1
50 - 51
3545
69
XM_017351033.2
XP_017206522.1
48 - 49
3415
69
XM_017351035.2
XP_017206524.1
48 - 49
3416
69
XM_017351036.2
XP_017206525.1
48 - 49
3416
69
XM_017351037.2
XP_017206526.1
48 - 49
3416
69
XM_017351038.2
XP_017206527.1
44 - 45
3106
69
XM_021481472.1
XP_021337147.1
49 - 50
3652
69
XM_021481473.1
XP_021337148.1
46 - 47
3277
69
XM_021481474.1
XP_021337149.1
41 - 42
3064
69
GenBank mRNA
DQ523562.1
-
51 - 52
3289
69
MF067533.1
-
11 - 12
617
69
MF067534.1
-
11 - 12
629
69
MF067535.1
-
11 - 12
622
69
MF067537.1
-
11 - 12
617
69
MF067538.1
-
12 - 13
659
69
Back To Top
More Information
Set Membership:
Amplicon length less than or equal to 70
Danio rerio
Probe spans exons
Panther Classification:
Molecular Function -
structural protein
Gene Ontology Categories:
Function(s)
Process(es)
biological_process
extracellular matrix structural constituent
Back To Top
Related Products
TaqMan
®
Fast Advanced Master Mix
SuperScript
®
VILO
™
cDNA Synthesis Kit
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7487cff77c-86c4h:80/10.239.166.160:80.
git-commit: c531a7a49d92b854cb004d8acd820c41186dcba6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.2-2024.09.38-1.0