Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: mpp2a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001327790.1 | NP_001314719.1 | 10 - 11 | 1284 | 67 | |
XM_021470936.1 | XP_021326611.1 | 10 - 11 | 1166 | 67 | ||
GenBank mRNA | BC152524.1 | - | 7 - 8 | 562 | 67 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Danio rerio Probe spans exons |