Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: alcama | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_131000.1 | NP_571075.1 | 11 - 12 | 1496 | 65 | |
XM_005172635.4 | XP_005172692.1 | 11 - 12 | 1522 | 65 | ||
GenBank mRNA | BC050482.1 | - | 11 - 12 | 1432 | 65 | |
BC071276.1 | - | 11 - 12 | 1497 | 65 | ||
CU458879.1 | - | 11 - 12 | 1337 | 65 | ||
CU458962.1 | - | 11 - 12 | 1443 | 65 | ||
L25057.1 | - | 1 - 2 | 146 | 65 | ||
L25273.1 | - | 11 - 12 | 1496 | 65 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Danio rerio Probe spans exons |