Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: lmnb2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_131002.2 | NP_571077.2 | 1 - 2 | 415 | 71 | |
GenBank mRNA | AJ005936.1 | - | 1 - 2 | 409 | 71 | |
BC162105.1 | - | 1 - 2 | 269 | 71 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Danio rerio Probe spans exons |