Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: lmnb2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_131002.2 | NP_571077.2 | 5 - 6 | 1005 | 92 | |
GenBank mRNA | AJ005936.1 | - | 5 - 6 | 999 | 92 | |
BC162105.1 | - | 5 - 6 | 859 | 92 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Danio rerio Probe spans exons |