Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: lmnb2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_131002.2 | NP_571077.2 | 9 - 10 | 1694 | 68 | |
GenBank mRNA | AJ005936.1 | - | 9 - 10 | 1688 | 68 | |
BC162105.1 | - | 9 - 10 | 1548 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |