Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: anos1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_131358.1 | NP_571433.1 | 4 - 5 | 558 | 61 | |
GenBank mRNA | AF163310.1 | - | 4 - 5 | 558 | 61 | |
BC060938.1 | - | 4 - 5 | 562 | 61 | ||
KM655630.1 | - | 4 - 5 | 511 | 61 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Danio rerio Probe spans exons |