Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: anos1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_131358.1 | NP_571433.1 | 6 - 7 | 880 | 85 | |
GenBank mRNA | AF163310.1 | - | 6 - 7 | 880 | 85 | |
BC060938.1 | - | 6 - 7 | 884 | 85 | ||
KM655630.1 | - | 6 - 7 | 833 | 85 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Danio rerio Probe spans exons |