Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: anos1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_131358.1 | NP_571433.1 | 11 - 12 | 1595 | 91 | |
GenBank mRNA | AF163310.1 | - | 11 - 12 | 1595 | 91 | |
BC060938.1 | - | 11 - 12 | 1599 | 91 | ||
KM655630.1 | - | 11 - 12 | 1548 | 91 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Danio rerio Probe spans exons |