Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: st8sia2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_153662.1 | NP_705948.1 | 5 - 6 | 1469 | 74 | |
GenBank mRNA | AJ705101.1 | - | 5 - 6 | 859 | 74 | |
AY055462.1 | - | 5 - 6 | 1469 | 74 | ||
BC050483.1 | - | 5 - 6 | 1422 | 74 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Danio rerio Probe spans exons |