Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: mib1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_173286.3 | NP_775393.2 | 1 - 2 | 330 | 103 | |
XM_009296844.3 | XP_009295119.1 | 1 - 2 | 1232 | 103 | ||
XM_009296845.3 | XP_009295120.1 | 1 - 2 | 1232 | 103 | ||
XM_009296846.3 | XP_009295121.1 | 1 - 2 | 1232 | 103 | ||
XM_009296848.3 | XP_009295123.1 | 1 - 2 | 1232 | 103 | ||
XM_009296854.3 | XP_009295129.1 | 1 - 2 | 1232 | 103 | ||
XM_009296859.3 | XP_009295134.1 | 1 - 2 | 1232 | 103 | ||
GenBank mRNA | AF506233.1 | - | 1 - 2 | 991 | 103 | |
AF537301.1 | - | 1 - 2 | 330 | 103 | ||
BC162476.1 | - | 1 - 2 | 290 | 103 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Danio rerio Probe spans exons |