Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: slc34a2b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_182877.2 | NP_878297.1 | 7 - 8 | 877 | 116 | |
XM_021469757.1 | XP_021325432.1 | 8 - 9 | 1327 | 116 | ||
GenBank mRNA | AF297180.2 | - | 7 - 8 | 878 | 116 | |
BC054940.1 | - | 8 - 9 | 896 | 116 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Danio rerio Probe spans exons |