Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ldlrap1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_198980.1 | NP_945331.1 | 2 - 3 | 319 | 65 | |
XM_005159645.3 | XP_005159702.1 | 2 - 3 | 371 | 65 | ||
GenBank mRNA | BC045926.1 | - | 2 - 3 | 319 | 65 | |
BC164648.1 | - | 2 - 3 | 264 | 65 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Danio rerio Probe spans exons |