Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: hacd2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_199861.1 | NP_956155.1 | 3 - 4 | 320 | 65 | |
GenBank mRNA | BC044396.1 | - | 3 - 4 | 320 | 65 | |
BC165084.1 | - | 3 - 4 | 282 | 65 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Danio rerio Probe spans exons |