Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: fam49ba | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001291713.1 | NP_001278642.1 | 3 - 4 | 592 | 61 | |
NM_200070.3 | NP_956364.2 | 3 - 4 | 592 | 61 | ||
GenBank mRNA | AY394961.1 | - | 3 - 4 | 292 | 61 | |
BC053261.1 | - | 3 - 4 | 278 | 61 | ||
EH541135.1 | - | 3 - 4 | 615 | 61 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |