Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: snrnp40 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_200322.1 | NP_956616.1 | 4 - 5 | 612 | 57 | |
GenBank mRNA | BC051783.1 | - | 4 - 5 | 612 | 57 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Danio rerio Probe spans exons |