Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: hyi | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_200334.1 | NP_956628.1 | 6 - 7 | 742 | 89 | |
GenBank mRNA | BC053111.1 | - | 6 - 7 | 742 | 89 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Danio rerio Probe spans exons |