Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: hsd17b3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_200364.1 | NP_956658.1 | 3 - 4 | 389 | 83 | |
XM_005167148.4 | XP_005167205.1 | 4 - 5 | 2034 | 83 | ||
GenBank mRNA | BC053203.1 | - | 3 - 4 | 389 | 83 | |
BC165921.1 | - | 2 - 3 | 210 | 83 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |