Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: rer1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_200675.2 | NP_956969.1 | 4 - 5 | 326 | 70 | |
XM_005167296.4 | XP_005167353.2 | 4 - 5 | 408 | 70 | ||
XM_005167298.4 | XP_005167355.2 | 4 - 5 | 393 | 70 | ||
XM_005167299.4 | XP_005167356.1 | 4 - 5 | 413 | 70 | ||
GenBank mRNA | AY398406.1 | - | 4 - 5 | 326 | 70 | |
BC050162.1 | - | 4 - 5 | 418 | 70 | ||
BC058292.1 | - | 4 - 5 | 328 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |