Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ddx39b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_200943.3 | NP_957237.1 | 8 - 9 | 1076 | 99 | |
GenBank mRNA | BC055240.1 | - | 8 - 9 | 1047 | 99 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |