Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: lmo4b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_212689.2 | NP_997854.2 | 2 - 3 | 608 | 66 | |
GenBank mRNA | AF398515.2 | - | 2 - 3 | 564 | 66 | |
BC076053.1 | - | 2 - 3 | 582 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Danio rerio Probe spans exons |