Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: prg4a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_212753.2 | NP_997918.2 | 5 - 6 | 431 | 68 | |
XM_005163364.4 | XP_005163421.1 | 5 - 6 | 454 | 68 | ||
GenBank mRNA | BC050178.1 | - | 5 - 6 | 431 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Danio rerio Probe spans exons |