Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: mpx | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_212779.2 | NP_997944.2 | 15 - 16 | 2342 | 110 | |
GenBank mRNA | AF349034.1 | - | 15 - 16 | 2343 | 110 | |
AF378824.1 | - | 12 - 13 | 2019 | 110 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Danio rerio Probe spans exons |