Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: aip | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_214712.1 | NP_999877.1 | 2 - 3 | 289 | 112 | |
GenBank mRNA | AF274877.1 | - | 2 - 3 | 289 | 112 | |
BC078231.1 | - | 2 - 3 | 313 | 112 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Danio rerio Probe spans exons |