Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: tbx2a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001102384.1 | NP_001095854.1 | 4 - 5 | 1252 | 97 | |
GenBank mRNA | BC068364.1 | - | 4 - 5 | 1224 | 97 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |