Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: rcn3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001002158.1 | NP_001002158.1 | 6 - 7 | 791 | 84 | |
XM_005164012.4 | XP_005164069.1 | 6 - 7 | 859 | 84 | ||
GenBank mRNA | BC071338.1 | - | 6 - 7 | 791 | 84 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |