Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: gmppaa | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001002196.1 | NP_001002196.1 | 11 - 12 | 1305 | 81 | |
XM_005167476.4 | XP_005167533.1 | 12 - 13 | 1451 | 81 | ||
GenBank mRNA | BC074036.1 | - | 11 - 12 | 1305 | 81 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |