Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: gmppaa | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001002196.1 | NP_001002196.1 | 7 - 8 | 892 | 85 | |
XM_005167476.4 | XP_005167533.1 | 8 - 9 | 1038 | 85 | ||
GenBank mRNA | BC074036.1 | - | 7 - 8 | 892 | 85 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |