Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: spag6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001002210.1 | NP_001002210.1 | 6 - 7 | 967 | 93 | |
GenBank mRNA | BC074072.1 | - | 6 - 7 | 967 | 93 | |
BC165187.1 | - | 6 - 7 | 887 | 93 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Danio rerio Probe spans exons |