Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: spag6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001002210.1 | NP_001002210.1 | 7 - 8 | 1123 | 71 | |
GenBank mRNA | BC074072.1 | - | 7 - 8 | 1123 | 71 | |
BC165187.1 | - | 7 - 8 | 1043 | 71 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |