Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: rpp21 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001003530.2 | NP_001003530.1 | 2 - 3 | 246 | 87 | |
GenBank mRNA | BC078300.1 | - | 2 - 3 | 250 | 87 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |