Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: lhx8a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001003980.2 | NP_001003980.1 | 4 - 5 | 670 | 72 | |
XM_005171154.4 | XP_005171211.1 | 4 - 5 | 72 | |||
GenBank mRNA | AY664404.1 | - | 4 - 5 | 671 | 72 | |
BC117601.1 | - | 4 - 5 | 72 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |