Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: quo | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001004109.2 | NP_001004109.2 | 15 - 16 | 4971 | 93 | |
GenBank mRNA | AY654285.2 | - | 15 - 16 | 4976 | 93 | |
BC162633.1 | - | 15 - 16 | 4897 | 93 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Danio rerio Probe spans exons |