Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: quo | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001004109.2 | NP_001004109.2 | 9 - 10 | 3007 | 136 | |
GenBank mRNA | AY654285.2 | - | 9 - 10 | 3006 | 136 | |
BC162633.1 | - | 9 - 10 | 2927 | 136 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Danio rerio Probe spans exons |