Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: atad1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001004640.3 | NP_001004640.1 | 3 - 4 | 409 | 69 | |
GenBank mRNA | BC081379.1 | - | 3 - 4 | 408 | 69 | |
BC164811.1 | - | 2 - 3 | 305 | 69 | ||
EH554125.1 | - | 3 - 4 | 444 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |