Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: cryba1l2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001005965.2 | NP_001005965.2 | 4 - 5 | 483 | 76 | |
GenBank mRNA | BC083301.1 | - | 4 - 5 | 475 | 76 | |
BC164982.1 | - | 3 - 4 | 318 | 76 | ||
EU935589.1 | - | 4 - 5 | 483 | 76 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Recommended Assay Danio rerio Probe spans exons |