Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: chga | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001006059.2 | NP_001006059.2 | 6 - 7 | 839 | 90 | |
GenBank mRNA | BC083265.1 | - | 6 - 7 | 825 | 93 | |
BC164760.1 | - | 6 - 7 | 673 | 93 | ||
CK027054.1 | - | 3 - 4 | 405 | 90 | ||
KM655711.1 | - | 6 - 7 | 666 | 93 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Danio rerio Probe spans exons |