Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: chga | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001006059.2 | NP_001006059.2 | 7 - 8 | 1207 | 72 | |
XM_005160561.4 | XP_005160618.1 | 7 - 8 | 1225 | 72 | ||
GenBank mRNA | BC083265.1 | - | 7 - 8 | 1193 | 72 | |
BC164760.1 | - | 7 - 8 | 1041 | 72 | ||
EH463078.1 | - | 1 - 2 | 549 | 72 | ||
KM655711.1 | - | 7 - 8 | 1034 | 72 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Danio rerio Probe spans exons |