Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: fyna | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001328163.1 | NP_001315092.1 | 7 - 8 | 963 | 92 | |
GenBank mRNA | AJ620748.1 | - | 7 - 8 | 979 | 92 | |
AY948197.1 | - | 7 - 8 | 949 | 92 | ||
BC075763.1 | - | 7 - 8 | 954 | 92 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Recommended Assay Danio rerio Probe spans exons |