Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: stx17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001007449.1 | NP_001007450.1 | 5 - 6 | 706 | 96 | |
XM_005158053.4 | XP_005158110.1 | 5 - 6 | 693 | 96 | ||
GenBank mRNA | BC085390.1 | - | 5 - 6 | 706 | 96 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |