Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: pinx1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001013265.2 | NP_001013283.2 | 3 - 4 | 233 | 136 | |
XM_021468369.1 | XP_021324044.1 | 4 - 5 | 326 | 136 | ||
XM_021468370.1 | XP_021324045.1 | 4 - 5 | 295 | 136 | ||
GenBank mRNA | AY576808.2 | - | 3 - 4 | 233 | 136 | |
BC054670.1 | - | 3 - 4 | 233 | 136 | ||
BC065317.1 | - | 2 - 3 | 209 | 136 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |