Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: gnpda1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001017867.1 | NP_001017867.1 | 4 - 5 | 481 | 75 | |
GenBank mRNA | BC092982.1 | - | 4 - 5 | 481 | 75 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |